4-Tolualdehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Methylbenzaldehyd
, p-Tolualdehyde
, 4-Tolylaldehyde
, p-Methylbenzaldehyd
, 4-Methylbenzaldehyde
, p-Tolylaldehyde
13900-1000
13900-0025
13900-5000
TS13900-0025EA
349.86
USD
TS13900-0025
TS13900-5000
TS13900-1000
4-Tolualdehyde 97%
4-Tolualdehyde
Formula:
C₈H₈O MW: 120.15 g/mol Boiling Pt: 204…205 °C (760 mmHg) Melting Pt: –6 °C Density: 1.015 g/cm³ Flash Pt: 71 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006954 CAS Number: 104-87-0 EINECS: 203-246-9 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5440 to 1.5480 (20°C, 589 nm) |
GC | ≥96.0 % |
Specific gravity | (20°C) 1.010 to 1.025 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...