Phenyl dichlorophosphate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenyl phosphorodichloridate
13048-1000
13048-0250
TS13048-1000EA
75
USD
TS13048-1000
TS13048-0250
Phenyl dichlorophosphate 99%
Phenyl dichlorophosphate
Formula:
C₆H₅Cl₂O₂P Boiling Pt: 241…243 °C (760 mmHg) Melting Pt: –1 °C Density: 1.412 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002067 CAS Number: 770-12-7 EINECS: 212-220-6 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5210 to 1.5230 (20°C, 589 nm) |
GC | ≥98.5 % |
Residue after evaporation | ≤0.002 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...