4-(2-Chloroethyl)anisole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-(2-Chloroethyl)-4-methoxybenzene

12311-1000 12311-0050
TS12311-1000EA 140.76 USD
TS12311-1000 TS12311-0050
4-(2-Chloroethyl)anisole 97%
4-(2-Chloroethyl)anisole
Formula: C₉H₁₁ClO
Boiling Pt: 125…128 °C (13mbar)
Density: 1.13 g/cm³
Flash Pt: 122 °C
Storage Temperature: Ambient
MDL Number: MFCD00044718
CAS Number: 18217-00-0
EINECS: 242-099-5

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5360 to 1.5380 (20°C, 589 nm)
GC ≥96.0 %
Specific gravity (20°C) 1.12 to 1.13

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR