5-Hydroxy-1,4-naphthoquinone (juglone) 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Hydroxy-1,4-naphthalenedione5-Hydroxy-p-naphthoquinone

12164-0050 12164-0010
TS12164-0050EA 173.4 USD
TS12164-0050 TS12164-0010
5-Hydroxy-1,4-naphthoquinone (juglone) 97%
5-Hydroxy-1,4-naphthoquinone (Juglone)
Formula: C₁₀H₆O₃
MW: 174.16 g/mol
Melting Pt: 161…163 °C
Storage Temperature: Ambient
MDL Number: MFCD00001684
EINECS: 207-567-5
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Orange to brown Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR