2,2'-Dithiodi(benzoic acid) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-Dithiodibenzoic acid2,2'-Dithiosalicylic acid

11756-5000 11756-1000
TS11756-5000EA 122.93 USD
TS11756-5000 TS11756-1000
2,2'-Dithiodi(benzoic acid) 98%
2,2'-Dithiodi(benzoic acid)
Formula: C₁₄H₁₀O₄S₂
MW: 306.36 g/mol
Melting Pt: 287…290 °C
Storage Temperature: Ambient
MDL Number: MFCD00002465
CAS Number: 119-80-2
EINECS: 204-352-8

Order Now

Specification Test Results

Appearance Brown Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Chloride (Cl) ≤5 ppm
Nitrate (NO3) ≤0.002 %
Ammonium (NH4) ≤0.002 %
Calcium (Ca) ≤0.02 %
Iron (Fe) ≤5 ppm
Manganese (Mn) ≤5 ppm
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR