4-Chloro-3-nitrobenzotrifluoride 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Chloro-3-nitrobenzotrifluoride4-Chloro-α-α-α-trifluoro-3-nitrotoluene1-Chloro-2-nitro-4-(trifluoromethyl)benzene

10970-1000
TS10970-1000EA 78 USD
TS10970-1000
4-Chloro-3-nitrobenzotrifluoride 98%
4-Chloro-3-nitrobenzotrifluoride
Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: –2 °C
Density: 1.511 g/cm³
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00007084
CAS Number: 121-17-5
EINECS: 204-451-6
UN: 2307
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4880 to 1.4910 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR