4-Chloro-α,α,α-trifluorotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Chlorobenzotrifluoride
, 1-Chloro-4-(trifluoromethyl)benzene
10873-0010
10873-2500
10873-0250
TS10873-0250EA
20.2
USD
TS10873-0250
TS10873-0010
TS10873-2500
4-Chloro-α,α,α-trifluorotoluene 98%
4-Chloro-α,α,α-trifluorotoluene
Formula:
C₇H₄ClF₃ MW: 180.56 g/mol Boiling Pt: 136…138 °C (760 mmHg) Melting Pt: –36 °C Density: 1.35 g/cm³ Flash Pt: 43 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000627 CAS Number: 98-56-6 EINECS: 202-681-1 UN: 2234 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4453 to 1.4473 (20°C, 589 nm) |
GC | ≥97.5 % |
Stabilizer | copper |
Acidity (CH3COOH) | ≤0.002 % |
Peroxides | ≤0.005 % (as H2O2) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...