sec-Butylbenzene 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Phenylbutane

10786-0050 10786-2500 10786-0500
TS10786-2500EA 130.56 USD
TS10786-2500 TS10786-0050 TS10786-0500
sec-Butylbenzene 99+%
sec-Butylbenzene
Formula: C₆H₅CH(CH₃)CH₂CH₃
MW: 134.22 g/mol
Boiling Pt: 173 °C (1013 hPa)
Melting Pt: –75 °C
Density: 0.86 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00009329
CAS Number: 135-98-8
EINECS: 205-227-0
UN: 2709
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/58/12/765812.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR