2-Bromomesitylene 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromomesitylene2-Bromo-1,3,5-trimethylbenzeneMesityl bromide2,4,6-Trimethylbromobenzene

10712-1000
TS10712-1000EA 101.59 USD
TS10712-1000
2-Bromomesitylene 99%
2-Bromomesitylene
Formula: C₉H₁₁Br
MW: 199.09 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.3 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00000073
CAS Number: 576-83-0
EINECS: 209-405-9

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5510 to 1.5530 (20 °C, 589 nm)
GC ≥98.5 %
Appearance of solution (0.5mmol/ml DMF) clear, colourless
Specific optical rotation +80.0° to +89.0° (20 °C, 589 nm) (c=1, chloroform) <0.2% D-enantiomer
Residual solvents ≤5 % (2-Propanol, THF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR