Dibutyl succinate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Succinic acid dibutyl ester
45301-1000
45301-0250
TS45301-1000EA
187
USD
TS45301-1000
TS45301-0250
Dibutyl succinate 99%
Dibutyl succinate
Formula:
C₁₂H₂₂O₄ MW: 230.3 g/mol Boiling Pt: 274 °C Melting Pt: –29 °C Density: 0.98 g/cm³ Flash Pt: 144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040495 CAS Number: 141-03-7 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4280 to 1.4310 (20 °C, 589 nm) |
GC | ≥98.5 % |
Acid value | ≤2.0 mg KOH/g |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...