3-Fluoro-4-formylpyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Fluoro-4-pyridinecarbaldehyde
, 3-Fluoro-4-pyridinecarboxaldehyde
, 3-Fluoroisonicotinaldehyde
44217-0050
44217-0010
TS44217-0050EA
344.33
USD
TS44217-0050
TS44217-0010
3-Fluoro-4-formylpyridine 97%
3-Fluoro-4-formylpyridine
Formula:
C₆H₄FNO MW: 125.1 g/mol Boiling Pt: 70 °C (25Torr) Density: 1.255 g/cm³ Flash Pt: 67 °C Storage Temperature: Ambient |
MDL Number:
MFCD06410686 CAS Number: 40273-47-0 |
Specification Test Results
Appearance | Clear colorless to orange to brown Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...