2-(2,2,2-Trifluoroacetyl)thiophene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Trifluoroacetyl)thiophene

43470-0050
TS43470-0050EA 119.36 USD
TS43470-0050
2-(2,2,2-Trifluoroacetyl)thiophene 98%
2-(2,2,2-Trifluoroacetyl)thiophene
Formula: C₆H₅ClOS
MW: 180.15
Flash Pt: 59 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041427
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC ≥98.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR