4-Hydroxy-3-methylbenzaldehyde 98%

Supplier: Thermo Scientific Chemicals
Warning

42755-0010
TS42755-0010EA 97.13 USD
TS42755-0010
4-Hydroxy-3-methylbenzaldehyde 98%
4-Hydroxy-3-methylbenzaldehyde
Formula: C₈H₈O₂
MW: 136.1479 g/mol
Melting Pt: 118…120 °C
Storage Temperature: Ambient
MDL Number: MFCD00012360
CAS Number: 15174-69-3

Order Now

Specification Test Results

Appearance Off-white to tan to brown Crystalline powder
Infrared spectrum Conforms
Melting point 118 to 120 °C
GC ≥97.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR