2-Hydroxyacetophenone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxyacetophenoneα-HydroxyacetophenonePhenacyl alcoholBenzoylcarbinol2-Hydroxy-1-phenylethanonePhenacyl hydroxyide

41190-0250
TS41190-0250EA 188.54 USD
TS41190-0250
2-Hydroxyacetophenone 97%
2-Hydroxyacetophenone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 118…120 °C (11 mmHg)
Melting Pt: 83…88 °C
MDL Number: MFCD00041829
CAS Number: 582-24-1
EINECS: 209-480-8

Order Now

Specification Test Results

Appearance Light yellow to orange Powder
Infrared spectrum Conforms
Melting point 83 to 88 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR