Ethyl trifluoropyruvate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 3,3,3-trifluoro-2-oxopropanoateEthyl 3,3,3-trifluoropyruvate

39901-0050
TS39901-0050EA 56 USD
TS39901-0050
Ethyl trifluoropyruvate 98+%
Ethyl trifluoropyruvate
Formula: C₅H₅F₃O₃
MW: 170.09
Flash Pt: 27 °C
Storage Temperature: Ambient
UN: 3272
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3390 to 1.3410 (20 °C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR