3-Formyl-6-methoxypyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Methoxy-3-pyridinecarboxaldehyde2-Methoxypyridine-5-carboxaldehyde6-Methoxynicotinaldehyde6-Methoxypyridine-3-carboxaldehyde

38057-0050 38057-0010
TS38057-0010EA 73 USD
TS38057-0010 TS38057-0050
3-Formyl-6-methoxypyridine 97%
3-Formyl-6-methoxypyridine
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 46…54 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02683446
CAS Number: 65873-72-5

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 46 to 54 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR