Methyl trifluoropyruvate 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Methyl 3,3,3-trifluoro-2-oxopropanoateMethyl 3,3,3-trifluoropyruvate

37971-0050
TS37971-0050EA 58 USD
TS37971-0050
Methyl trifluoropyruvate 97%
Methyl trifluoropyruvate
Formula: C₄H₃F₃O₃
MW: 156.06 g/mol
Flash Pt: 26 °C
Storage Temperature: Ambient
UN: 2924
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3320 to 1.3340 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR