tert-Butyl ethyl malonate 97%

Supplier: Thermo Scientific Chemicals

33940-0250 33940-0010
TS33940-0250EA 90.89 USD
TS33940-0250 TS33940-0010
tert-Butyl ethyl malonate 97%
tert-Butyl ethyl malonate
Formula: C₉H₁₆O₄
Boiling Pt: 106…110 °C (760 mmHg)
Density: 1.001 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00009193
EINECS: 251-269-8

Order Now

Specification Test Results

Appearance Clear colorless to pale yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR