Ethyl-3-phenylglycidate (mixture of cis and trans isomers) cis- and trans-mixture 90%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 3-phenylglycidate
33265-0500
33265-0050
33265-2500
TS33265-0500EA
45
USD
TS33265-0500
TS33265-0050
TS33265-2500
Ethyl-3-phenylglycidate (mixture of cis and trans isomers) cis- and trans-mixture 90%
Ethyl 3-phenylglycidate (mixture of cis and trans isomers)
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Boiling Pt: 96 °C (.5 mmHg) Density: 1.12 g/cm³ Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005123 CAS Number: 121-39-1 EINECS: 204-467-3 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5165 to 1.5205 (20 °C, 589 nm) |
GC | ≥88 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...