3-Bromo-1,1,1-trifluoroacetone 97%

Supplier: Thermo Scientific Chemicals
Danger

29744-0050
TS29744-0050EA 50 USD
TS29744-0050
3-Bromo-1,1,1-trifluoroacetone 97%
3-Bromo-1,1,1-trifluoroacetone
Formula: BrH₂CC(O)CF₃
MW: 190.95 g/mol
Boiling Pt: 87 °C (1013 hPa)
Density: 1.83 g/cm³
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00039237
CAS Number: 431-35-6
EINECS: 207-071-9
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3740 to 1.3770 (20 °C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR