4-Phenylbutyrophenone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1,4-Diphenyl-1-butanone

23282-0010
TS23282-0010EA 43 USD
TS23282-0010
4-Phenylbutyrophenone 98%
4-Phenylbutyrophenone
Formula: (C₆H₅)C(O)CH₂CH₂CH₂(C₆H₅)
MW: 224.3 g/mol
Boiling Pt: 155 °C (0.2 mmHg)
Melting Pt: 55…59 °C
MDL Number: MFCD00022005
CAS Number: 5407-91-0
EINECS: 226-471-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR