1-Heptadecanol 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Heptadecan-1-olHexadecyl carbinol

22123-0250 22123-0050
TS22123-0250EA 210 USD
TS22123-0250 TS22123-0050
1-Heptadecanol 97%
1-Heptadecanol
Formula: H₃C(CH₂)₁₅CH₂OH
MW: 256.47 g/mol
Boiling Pt: 308.5 °C (760 mmHg)
Melting Pt: 51…56 °C
Flash Pt: 154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002822
CAS Number: 1454-85-9
EINECS: 215-932-5

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 51 to 56 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR