Ethyl malonyl chloride 90% tech.

Supplier: Thermo Scientific Chemicals
Danger

17269-0050 17269-1000 17269-0250
TS17269-0250EA 176.3 USD
TS17269-0250 TS17269-0050 TS17269-1000
Ethyl malonyl chloride 90% tech.
Ethyl malonyl chloride
Formula: C₅H₇ClO₃
Boiling Pt: 79…80 °C (25 mmHg)
Density: 1.17 g/cm³
Flash Pt: 67 °C
Storage Temperature: Ambient
MDL Number: MFCD00000736
EINECS: 252-934-5
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4250 to 1.4310 (20 °C, 589 nm)
Titration RCOCl ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR