Cyclobutanol 99+%

Supplier: Thermo Scientific Chemicals
Danger

16875-0050 16875-2500 16875-0010
TS16875-0010EA 109 USD
TS16875-0010 TS16875-2500 TS16875-0050
Cyclobutanol 99+%
Cyclobutanol
Formula: C₄H₈O
Boiling Pt: 123…124 °C (733 mmHg)
Density: 0.921 g/cm³
Flash Pt: 21 °C
Storage Temperature: Ambient
MDL Number: MFCD00001318
CAS Number: 2919-23-5
EINECS: 220-858-1
UN: 1987
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4370 (20 °C, 589 nm)
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR