3,5-Di-tert-butyl-4-hydroxybenzaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

16121-0250
TS16121-0250EA 79.43 USD
TS16121-0250
3,5-Di-tert-butyl-4-hydroxybenzaldehyde 97%
3,5-Di-tert-butyl-4-hydroxybenzaldehyde
Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Boiling Pt: 332 °C (1013 hPa)
Melting Pt: 186…190 °C
MDL Number: MFCD00008826
CAS Number: 1620-98-0
EINECS: 216-592-0

Order Now

Specification Test Results

Appearance Light yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 190 °C
GC ≥96.0 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR