Propiophenone 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl phenyl ketonePhenyl ethyl ketone1-Phenyl-1-propanone

13154-2500 13154-0020 13154-0025 13154-0010
TS13154-0025EA 158.78 USD
TS13154-0025 TS13154-2500 TS13154-0010 TS13154-0020
Propiophenone 99%
Propiophenone
Formula: C₆H₅C(O)CH₂CH₃
MW: 134.18 g/mol
Boiling Pt: 218 °C (760 mmHg)
Melting Pt: 18 °C
Density: 1 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00009309
CAS Number: 93-55-0
EINECS: 202-257-6

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5250 to 1.5280 (20 °C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR