2-Methoxypyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

12610-0050 12610-1000 12610-0250
TS12610-1000EA 87.82 USD
TS12610-1000 TS12610-0250 TS12610-0050
2-Methoxypyridine 98%
2-Methoxypyridine
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 142 °C (1013 hPa)
Density: 1.04 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00006262
CAS Number: 1628-89-3
EINECS: 216-623-8
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5030 to 1.5050 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR