Menthone (mixture of isomers) 98%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
2-Isopropyl-5-Methylcyclohexanone
12541-1000
TS12541-1000EA
131.56
USD
TS12541-1000
Menthone (mixture of isomers) 98%
Menthone (mixture of isomers)
Formula:
C₁₀H₁₈O MW: 154.25 g/mol Melting Pt: –6 °C Density: 0.89 g/cm³ Flash Pt: 69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00062998 CAS Number: 10458-14-7 EINECS: 233-944-9 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4460 to 1.4550 (20 °C, 589 nm) |
Density | (20°C) 0.889 to 0.898 g/ml |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...