Elaidic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

11773-0050 11773-0010
TS11773-0050EA 169.73 USD
TS11773-0050 TS11773-0010
Elaidic acid 98%
Elaidic acid
Formula: C₁₈H₃₄O₂
Boiling Pt: 288 °C (100 mmHg)
Melting Pt: 43…45 °C
Flash Pt: >110 °C
Storage Temperature: Freezer
MDL Number: MFCD00063954
CAS Number: 112-79-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 42 to 44 °C
GC ≥97.5 %
Chloride (Cl) ≤0.5 ppm
Sulfate (SO4) ≤1 ppm
Arsenic (As) ≤0.01 ppm
Iron (Fe) ≤0.2 ppm
Heavy metals (as Pb) ≤0.2 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR