Diisopropyl phosphonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diisopropyl phosphiteDiisopropyl hydrogen phosphiteDi-i-propylphosphite

11526-1000
TS11526-1000EA 99.86 USD
TS11526-1000
Diisopropyl phosphonate 98%
Diisopropyl phosphonate
Formula: C₆H₁₅O₃P
MW: 166.16 g/mol
Boiling Pt: 71 °C (8 mmHg)
Density: 0.997 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00117905
CAS Number: 1809-20-7
EINECS: 217-317-7

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4065 to 1.4085 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR