Diethyl allylmalonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11396-1000
11396-5000
TS11396-1000EA
201
USD
TS11396-1000
TS11396-5000
Diethyl allylmalonate 97%
Diethyl allylmalonate
Formula:
C₁₀H₁₆O₄ Density: 1.01 g/cm³ Flash Pt: 92 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009155 CAS Number: 2049-80-1 EINECS: 218-072-9 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4300 to 1.4320 (20 °C, 589 nm) |
GC | ≥96.0 % |
Enantiomeric excess | ≥97.5 % |
Specific optical rotation | +29° to +33° (20°C, 589 nm) (Neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...