7-Hydroxyflavanone 98%

Supplier: Thermo Scientific Chemicals
Warning

36417-0010
TS36417-0010EA 58 USD
TS36417-0010
7-Hydroxyflavanone 98%
7-Hydroxyflavanone

Order Now

Specification Test Results

Appearance Light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 189 to 197 °C
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR