Perfluoroheptanoic acid 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Perfluoroenanthic acidTridecafluoroheptylic acidTridecafluoroenanthic acidTridecafluoroheptanoic acid

27727-0100
TS27727-0100EA 102 USD
TS27727-0100
Perfluoroheptanoic acid 96%
Perfluoroheptanoic acid
Formula: C₇HF₁₃O₂
Melting Pt: 30 °C
Flash Pt: >110 °C
MDL Number: MFCD00039604
CAS Number: 375-85-9
EINECS: 206-798-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or solid
Infrared spectrum Conforms
Titration with NaOH ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR