Iodocyclohexane 98% stabilized

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Cyclohexyl iodide

21849-1000 21849-0250
TS21849-1000EA 271.15 USD
TS21849-1000 TS21849-0250
Iodocyclohexane 98% stabilized
Iodocyclohexane
Formula: C₆H₁₁I
MW: 210.06 g/mol
Boiling Pt: 80…81 °C (20 mmHg)
Density: 1.62 g/cm³
Flash Pt: 71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003826
CAS Number: 626-62-0
EINECS: 210-957-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5450 to 1.5490 (20°C, 589 nm)
GC ≥97.5 %
Stabilizer Copper

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR