1,4-Dibromobutane 99%

Supplier: Thermo Scientific Chemicals
Danger

16128-0010 16128-5000 16128-1000
TS16128-5000EA 173 USD
TS16128-5000 TS16128-0010 TS16128-1000
1,4-Dibromobutane 99%
1,4-Dibromobutane
Formula: BrH₂C(CH₂)₂CH₂Br
MW: 215.92 g/mol
Boiling Pt: 197 °C (760 mmHg)
Melting Pt: –20 °C
Density: 1.808 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000261
CAS Number: 110-52-1
EINECS: 203-775-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.5176 to 1.5196 (20°C, 589 nm)
GC ≥98.5 %
Specific optical rotation +18° to +23° (20°C, 589 nm) (c=0.5, acetone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR