2-Bromoethyl acetate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetic acid (2-bromoethyl) ester

16009-0250
TS16009-0250EA 77 USD
TS16009-0250
2-Bromoethyl acetate ≥97%
2-Bromoethyl acetate
Formula: C₄H₇BrO₂
MW: 167 g/mol
Boiling Pt: 159 °C (1013 hPa)
Melting Pt: –14 °C
Density: 1.4623 g/cm³ (20 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00000235
CAS Number: 927-68-4
UN: 3334
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR