Methylcyclopentadiene dimer 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15685-0010
15685-0025
15685-2500
TS15685-0025EA
454
USD
TS15685-0025
TS15685-2500
TS15685-0010
Methylcyclopentadiene dimer 95%
Methylcyclopentadiene dimer
Formula:
C₁₂H₁₆ Boiling Pt: 200 °C (760 mmHg) Melting Pt: –51 °C Density: 0.94 g/cm³ Flash Pt: 26 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082297 CAS Number: 26472-00-4 EINECS: 247-724-5 UN: 3295 ADR: 3,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
NMR (1H-NMR) | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...