2-(Ethylthio)ethanol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Ethylmercapto)-ethanolEthyl 2-hydroxyethyl sulfideEthyl 2-hydroxyethyl sulphide

11855-1000 11855-0050
TS11855-1000EA 46 USD
TS11855-1000 TS11855-0050
2-(Ethylthio)ethanol 98%
2-(Ethylthio)ethanol
Formula: C₄H₁₀OS
Melting Pt: –100 °C
Density: 1.02 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00002909
CAS Number: 110-77-0
EINECS: 203-802-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4859 to 1.4879 (20°C, 589 nm)
GC ≥97.5 %
Nitrate (NO3) ≤20 ppm
Ammonium (NH4) ≤10 ppm
Calcium (Ca) ≤50 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤30 ppm
Magnesium (Mg) ≤50 ppm
Manganese (Mn) ≤3 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤500 ppm
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR