2,6-Dichlorotoluene 99+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1,3-Dichloro-2-methylbenzene
11379-0025
11379-1000
11379-5000
TS11379-5000EA
232
USD
TS11379-5000
TS11379-0025
TS11379-1000
2,6-Dichlorotoluene 99+%
2,6-Dichlorotoluene
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 196…203 °C (760 mmHg) Melting Pt: 2 °C Density: 1.25 g/cm³ Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000576 CAS Number: 118-69-4 EINECS: 204-269-7 UN: 3082 ADR: 9,III |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5500 to 1.5520 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...