Dibenzyl phosphonate 90+%, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dibenzyl phosphite

11266-0250 11266-1000 11266-0050
TS11266-1000EA 313.84 USD
TS11266-1000 TS11266-0050 TS11266-0250
Dibenzyl phosphonate 90+%, Technical Grade
Dibenzyl phosphonate
Formula: C₁₄H₁₅O₃P
Boiling Pt: 110…120 °C (.1 mmHg)
Density: 1.18 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004774
CAS Number: 17176-77-1
EINECS: 241-226-1

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5520 to 1.5550 (20°C, 589 nm)
NMR ≥90.0 %
UV at 260 nm A: ≤0.05
UV at 280 nm A: ≤0.05
pH 5.5 to 8.0 (5 % in water)
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR