Cycloheptane 99%

Supplier: Thermo Scientific Chemicals
Danger

11101-1000 11101-0050 11101-0250
TS11101-1000EA 361.79 USD
TS11101-1000 TS11101-0250 TS11101-0050
Cycloheptane 99%
Cycloheptane
Formula: C₇H₁₄
Boiling Pt: 118.4 °C (760 mmHg)
Melting Pt: –12 °C
Density: 0.81 g/cm³
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004147
CAS Number: 291-64-5
EINECS: 206-030-2
UN: 2241
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4440 to 1.4460 (20°C, 589 nm)
GC ≥98.5 %
Acid value 230 to 260 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR