1-Bromonaphthalene 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
α-Bromonaphthalene
10715-5000
10715-1000
10715-0025
TS10715-5000EA
391
USD
TS10715-5000
TS10715-0025
TS10715-1000
1-Bromonaphthalene 96%
1-Bromonaphthalene
Formula:
C₁₀H₇Br MW: 207.07 g/mol Boiling Pt: 279…281 °C (760 mmHg) Melting Pt: –1 °C Density: 1.489 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003868 CAS Number: 90-11-9 EINECS: 201-965-2 |
Specification Test Results
Appearance | Clear yellow to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6560 to 1.6590 (20°C, 589 nm) |
GC | ≥95 % |
GC | ≤2.5 % 2-Bromonaphthalene |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...