1H-Indazole-7-carboxylic acid 0.97

Supplier: Thermo Scientific Chemicals
Warning

42960-0010
TS42960-0010EA 207.91 USD
TS42960-0010
1H-Indazole-7-carboxylic acid 0.97
1H-Indazole-7-carboxylic acid
Formula: C₈H₆N₂O₂
Melting Pt: 229…231 °C
Storage Temperature: Ambient
MDL Number: MFCD06804572
CAS Number: 677304-69-7

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 228 to 233 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR