1-Boc-4-(methylamino)piperidine 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Methylamino)piperidine, N1-BOC protectedtert-Butyl 4-(methylamino)piperidine-1-carboxylate

45480-0010
TS45480-0010EA 70 USD
TS45480-0010
1-Boc-4-(methylamino)piperidine 95%
1-Boc-4-(methylamino)piperidine
Formula: C₁₁H₂₂N₂O₂
MW: 214.3046 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD02259411
CAS Number: 147539-41-1

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR