(4-Methoxyphenyl)acetyl chloride 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methoxyphenylacetyl chloride

36893-0020 36893-0100
TS36893-0100EA 85 USD
TS36893-0100 TS36893-0020
(4-Methoxyphenyl)acetyl chloride 98%
(4-Methoxyphenyl)acetyl chloride
Formula: C₉H₉ClO₂
MW: 184.62 g/mol
Boiling Pt: 143 °C (10 mmHg)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075474
CAS Number: 4693-91-8
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow to red Liquid
Infrared spectrum Conforms
Refractive index 1.5390 to 1.5420 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR