5-Bromo-2,4-dimethoxypyrimidine 0.97

Supplier: Thermo Scientific Chemicals
Warning

44339-0050
TS44339-0050EA 285.8 USD
TS44339-0050
5-Bromo-2,4-dimethoxypyrimidine 0.97
5-Bromo-2,4-dimethoxypyrimidine
Formula: C₆H₇BrN₂O₂
Boiling Pt: 125 °C (17Torr)
Melting Pt: 63…65 °C
Storage Temperature: Ambient
MDL Number: MFCD00038016
CAS Number: 56686-16-9

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 62 to 67 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR