Calcium thioglycolate trihydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Calcium thioglycolate trihydrate
, Mercaptoacetic acid calcium salt trihydrate
, Calcium bis(mercaptoacetate) trihydrate
, Calcium mercaptoacetate trihydrate
36703-1000
36703-5000
TS36703-1000EA
32
USD
TS36703-1000
TS36703-5000
Calcium thioglycolate trihydrate 99%
Calcium bis(mercaptoacetate) (Calcium thioglycolate) trihydrate
Formula:
C₄H₆CaO₄S₂·3H₂O MW: 276.35 g/mol Melting Pt: 270…280 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150707 CAS Number: 65208-41-5 EINECS: 212-402-5 UN: 3263 |
Specification Test Results
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Iodometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...