Ethyl-3-bromopropionate 0.99

Supplier: Thermo Scientific Chemicals
Warning

40981-1000 40981-0250
TS40981-1000EA 205.86 USD
TS40981-1000 TS40981-0250
Ethyl-3-bromopropionate 0.99
Ethyl 3-bromopropionate
Formula: C₅H₉BrO₂
MW: 181.03 g/mol
Boiling Pt: 69…71 °C (10 mmHg)
Density: 1.419 g/cm³
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD00000251
CAS Number: 539-74-2
EINECS: 208-724-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4510 to 1.4540 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR