2-Bromo-3,3,3-trifluoropropene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Bromo-3,3,3-trifluoro-1-propene
43446-0010
43446-0050
TS43446-0010EA
52
USD
TS43446-0010
TS43446-0050
2-Bromo-3,3,3-trifluoropropene 97%
2-Bromo-3,3,3-trifluoropropene
Formula:
C₃H₂BrF₃ MW: 174.95 g/mol Boiling Pt: 34 °C (760 mmHg) Density: 1.65 g/cm³ Storage Temperature: Refrigerator |
MDL Number:
MFCD00077469 CAS Number: 1514-82-5 |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3520 to 1.3570 (20°C, 589 nm) |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...