(2-Bromoethyl)cyclohexane 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2-cyclohexylethane2-Cyclohexylethyl bromide

39264-0250 39264-0050
TS39264-0250EA 225 USD
TS39264-0250 TS39264-0050
(2-Bromoethyl)cyclohexane 98%
(2-Bromoethyl)cyclohexane
Formula: C₈H₁₅Br
MW: 191.11 g/mol
Boiling Pt: 209 °C (760 mmHg)
Melting Pt: –57 °C
Density: 1.221 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00019398
CAS Number: 1647-26-3

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4890 to 1.4910 (20°C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR